Learn More
World Precision Instrument Bare Platinum Wire-PTP401
SDP
Catalog No. 50822168
Encompass
Uncoated pure platinum wire (PTPxxx) is 99.95% pure.
Catalog No. 50-822-168 Supplier World Precision Instrument Supplier No. PTP401
Uncoated pure platinum wire (PTPxxx) is 99.95% pure.