Learn More
Protech International Inc Platinum Plate Glass Slide Electrode, 1mm Gap x 10mm Length
SDP
Catalog No. NC1879374
Encompass
Platinum plate electrode on tempered glass slide, can hold up to ~40 mouse zygotes in a 5 L electroporation buffer. This enables efficient and high throughput zygote genome editing using CRISPR/Cas9 by electroporation.
Catalog No. NC1879374 Supplier Protech International Inc Supplier No. LF501PT110
 Restricted Item
Add to Cart
Add to Cart
Platinum plate electrode on tempered glass slide, can hold up to ~40 mouse zygotes in a 5 L electroporation buffer. This enables efficient and high throughput zygote genome editing using CRISPR/Cas9 by electroporation.