Learn More
Strem, An Ascensus Company CAS# 7440-06-4. 25cm. Platinum wire (99.95%). MFCD00011179
SDP
Catalog No. 5090113193
Encompass
CAS# 7440-06-4. 25cm. Platinum wire (99.95%). MFCD00011179. Molecular Weight: 195.09. Molecular Formula: Pt. Color/form: 0.5mm dia. (~1.05g/25cm) Strem# 78-0085. http://www.strem.com/catalog/v/78-0085/
Catalog No. 50-901-13193 Supplier Strem, An Ascensus Company Supplier No. 78008525CM
CAS# 7440-06-4. 25cm. Platinum wire (99.95%). MFCD00011179. Molecular Weight: 195.09. Molecular Formula: Pt. Color/form: 0.5mm dia. (~1.05g/25cm) Strem# 78-0085. http://www.strem.com/catalog/v/78-0085/