Learn More
Sigma Aldrich Platinum Wire Auxiliary Electrode
Catalog No. BASMW1032 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BASMW1032 1 Ea.
1 options
Catalog No. BASMW1032 Supplier Sigma Aldrich Supplier No. BASMW10321EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Platinum Wire Auxiliary Electrode (7.5 cm), with gold-plated connector, mounted in CTFE cylinder.Product of Bioanalytical Systems, Inc.
Specifications
Quantity 1 Ea.