Learn More
Sigma Aldrich Fine Chemicals Biosciences Platinum wire diam. 2.0 mm7G
Catalog No. 502903401
Encompass
Platinum (Pt) wire is a metal wire that is a biocompatible material that can be used in electrodes and implants. The efficiency of the platinum electrode can be improved by forming a platinum black surface.
Catalog No. 50-290-3401 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4446857G
Platinum (Pt) wire is a metal wire that is a biocompatible material that can be used in electrodes and implants. The efficiency of the platinum electrode can be improved by forming a platinum black surface.