Learn More
Glass Expansion Platinum X-Skimmer Cone (Boron-Free) for Thermo Scientific Element 1/Element 2/Neptune ICP-MS (1142150)
SDP
Catalog No. 501907713
Encompass_Preferred
Platinum X-Skimmer Cone, Boron-Free (1142150) ¿ For Thermo/Finnigan Element 1, 2 & Neptune ICP-MS. Ideal for lowest boron background and semiconductor applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest copper background, we reccommend Nickel Skimmler, see catalog number 501907706.For high sensitivity applications we reccommend Nickel X-Skimmer with Copper Core, see catalog number 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7713 Supplier Glass Expansion Supplier No. TF1007XPT
Platinum X-Skimmer Cone, Boron-Free (1142150) ¿ For Thermo/Finnigan Element 1, 2 & Neptune ICP-MS. Ideal for lowest boron background and semiconductor applications, including corrosive samples (e.g., routine aqueous environmental samples, >5% acid matrix, HF, organic). Although the highest cost option, it is typically the most durable, longest-lasting, and because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For lowest copper background, we reccommend Nickel Skimmler, see catalog number 501907706.For high sensitivity applications we reccommend Nickel X-Skimmer with Copper Core, see catalog number 501907707.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001