Learn More
MSC Platt Clamshell Hard Case, 8-1/2 in Wide, 1 ea
Catalog No. 5000768211
Encompass
This black clamshell hard case is constructed from high-density polyethylene and features cubed foam for customizable storage and protection.
  • Electrical insulator
  • Crack resistant
  • Mechanical hinge system
  • Scuff resistant
  • Double-wall construction
  • High-density
  • Dent resistant
  • Textured exteriors
  • Economical
  • Strong
  • Meets U.S. 94B standards
Catalog No. 50-007-68211 Supplier MSC Supplier No. 65359101
This black clamshell hard case is constructed from high-density polyethylene and features cubed foam for customizable storage and protection.
  • Electrical insulator
  • Crack resistant
  • Mechanical hinge system
  • Scuff resistant
  • Double-wall construction
  • High-density
  • Dent resistant
  • Textured exteriors
  • Economical
  • Strong
  • Meets U.S. 94B standards