Learn More
Medchemexpress LLC Platycodin A | 66779-34-8 | 99.3% | 1267.36 | 1 MG
SDP
Catalog No. 5000425265
Encompass_Preferred
Platycodin A is a monoacetylated saponin compound isolated from the roots of Platycodon grandiflorum A. DC. It exhibits antiproliferative activity against human cell lines and is intended for research use only.
  • Monoacetylated saponin compound
  • Isolated from the roots of Platycodon grandiflorum A. DC.
  • Exhibits antiproliferative activity against human cell lines
  • Solid, white to off-white appearance
  • Recommended storage at -20°C, protected from light
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-25265 Supplier Medchemexpress LLC Supplier No. HYN89221MG
Add to Cart
Add to Cart
Platycodin A is a monoacetylated saponin compound isolated from the roots of Platycodon grandiflorum A. DC. It exhibits antiproliferative activity against human cell lines and is intended for research use only.
  • Monoacetylated saponin compound
  • Isolated from the roots of Platycodon grandiflorum A. DC.
  • Exhibits antiproliferative activity against human cell lines
  • Solid, white to off-white appearance
  • Recommended storage at -20°C, protected from light
  • In solvent, recommended storage at -80°C for 6 months or -20°C for 1 month, protected from light