Learn More
Apexbio Technology LLC PLATYCODIN-D-20MG
SDP
Catalog No. 5000567423
Encompass_Preferred
Platycodin D (CAS No 58479-68-8) is a triterpenoid saponin primarily isolated from the root of Platycodon grandiflorus It has been studied in the biomedical research category for its potential anticancer properties affecting pathways such as apoptosis and cell cycle regulation Platycodin D exhibits cytotoxicity in the nanomolar to low micromolar range demonstrating efficacy in various in vitro cell models It has also been used in animal studies and drug screening to explore its therapeutic potential Experimental concentrations typically depend on the specific research design Its mechanism involves modulating signaling pathways contributing to its therapeutic effects
Catalog No. 50-005-67423 Supplier Apexbio Technology LLC Supplier No. N149820
Platycodin D (CAS No 58479-68-8) is a triterpenoid saponin primarily isolated from the root of Platycodon grandiflorus It has been studied in the biomedical research category for its potential anticancer properties affecting pathways such as apoptosis and cell cycle regulation Platycodin D exhibits cytotoxicity in the nanomolar to low micromolar range demonstrating efficacy in various in vitro cell models It has also been used in animal studies and drug screening to explore its therapeutic potential Experimental concentrations typically depend on the specific research design Its mechanism involves modulating signaling pathways contributing to its therapeutic effects