Learn More
TARGETMOL CHEMICALS INC PLATYCODIN D 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T388925MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Platycodin D may stimulate TNF-(alpha) synthesis or inhibit degradation of TNF-(alpha) mRNA. Platycodin D (10/30 uM) suppresses prostaglandin E2 production in rat peritoneal macrophages stimulated by the protein kinase C activator 12-O-tetradecanoyl phorbol 13-acetate (TPA). Platycodin D and D3 are useful as expectorant agents in the treatment of various airway diseases they can increase mucin release from rat and hamster tracheal surface epithelial cell culture and also from intact rat trachea upon nebulization. purity: 99%
Catalog No. 50-258-0080