Learn More
TARGETMOL CHEMICALS INC Platycodin D2 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN20885MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Platycodin D2 , a less hemolytic saponin, can improve specific cellular and humoral responses to hepatitis B surface antigen in mice, it could be safely used as adjuvant eliciting Th1 and Th2 immune responses. Purity 98.83%
Catalog No. 50-287-7077