Learn More
Medchemexpress LLC HY-N4087 5mg Medchemexpress, Platycodin D2 CAS:66663-90-9 Purity:>98%
SDP
Catalog No. 501970418
Encompass_Preferred
Medchemexpress, HY-N4087 5mg Platycodin D2 CAS:66663-90-9 Platycodin D2 is a saponin isolated from Platycodon grandiflorum, with anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0418 Supplier Medchemexpress LLC Supplier No. HYN40875MG
Medchemexpress, HY-N4087 5mg Platycodin D2 CAS:66663-90-9 Platycodin D2 is a saponin isolated from Platycodon grandiflorum, with anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.