Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Platycodin D2 | 66663-90-9 | 10 MM * 1 ML
SDP

Catalog No. 5000418812
Encompass_Preferred

Platycodin D2 is a saponin isolated from Platycodon grandiflorum, known for its anti-cancer activity and intended for research use only.

  • High purity: 99.78%
  • Molecular weight: 1387.46
  • Appearance: Off-white to light yellow solid
  • Exhibits antiproliferative activity against human HCT-15 and MES-SA cells
  • Available as a 10 mM solution in DMSO

Catalog No. 50-004-18812 Supplier Medchemexpress LLC Supplier No. HYN408710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Platycodin D2 is a saponin isolated from Platycodon grandiflorum, known for its anti-cancer activity and intended for research use only.

  • High purity: 99.78%
  • Molecular weight: 1387.46
  • Appearance: Off-white to light yellow solid
  • Exhibits antiproliferative activity against human HCT-15 and MES-SA cells
  • Available as a 10 mM solution in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.