Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Platycodin D2 | 66663-90-9 | MFCD29904544 | 99.8% | 1387.46 g/mol | C63H102O33 | 10 MG
SDP

Catalog No. 5000289473
Encompass_Preferred

Platycodin D2 is a triterpenoid saponin isolated from Platycodon grandiflorum with reported anti-cancer activity. It is supplied as a high-purity research reagent with documented solubility and storage guidance for experimental use.

  • Naturally derived triterpenoid saponin
  • Reported anti-cancer activity in preclinical studies
  • High purity (99.78%) suitable for research applications
  • CAS number 66663-90-9 for unambiguous identification
  • Soluble in DMSO (100 mg/mL); in vivo formulation suggestions available
  • Powder stable at -20°C for long-term storage

Catalog No. 50-002-89473 Supplier Medchemexpress LLC Supplier No. HYN408710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Platycodin D2 is a triterpenoid saponin isolated from Platycodon grandiflorum with reported anti-cancer activity. It is supplied as a high-purity research reagent with documented solubility and storage guidance for experimental use.

  • Naturally derived triterpenoid saponin
  • Reported anti-cancer activity in preclinical studies
  • High purity (99.78%) suitable for research applications
  • CAS number 66663-90-9 for unambiguous identification
  • Soluble in DMSO (100 mg/mL); in vivo formulation suggestions available
  • Powder stable at -20°C for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.