Learn More
Medchemexpress LLC HY-N3519 1mg Medchemexpress, Platycodin D3 CAS:67884-03-1 Purity:>98%
SDP
Catalog No. 502082914
Encompass_Preferred
Medchemexpress, HY-N3519 1mg Platycodin D3 CAS:67884-03-1 Platycodin D3 is a triterpenoid saponin isolated from Platycodon grandiflorum, with anti-HCV activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2914 Supplier Medchemexpress LLC Supplier No. HYN35191MG
Medchemexpress, HY-N3519 1mg Platycodin D3 CAS:67884-03-1 Platycodin D3 is a triterpenoid saponin isolated from Platycodon grandiflorum, with anti-HCV activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.