Learn More
TARGETMOL CHEMICALS INC Platycodin D3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN20895MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg and bulk. Please contact Fisher for quotes. Platycodin D3 is a NF-kB inhibitor, it could as expectorants in diverse inflammatory pulmonary diseases, it can regulate the production and secretion of airway mucin; it can significantly enhance mitogen- and OVA-induced splenocyte proliferation in the OVA-immunized mice. Platycodin D3 also exerts anti-Hepatitis C virus (HCV) activity. Purity 99.69%
Catalog No. 50-287-6555