Learn More
Medchemexpress LLC Platyconic acid A | 68051-23-0 | 99.1% | 1239.31 | 5 MG
SDP
Catalog No. 5000426187
Encompass_Preferred
Platyconic Acid A is an active component of changkil saponins from platycodon grandiflorum and can be used for the research of reducing airway inflammation.
  • Inhibits PMA-induced NF-κB p65 phosphorylation, IκBα phosphorylation and degradation and Akt phosphorylation in a concentration-dependent manner.
  • Decreases PMA-increased MUC5AC mRNA expression in a concentration-dependent manner.
  • Stored at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in H2O at 25 mg/mL (20.17 mM) with ultrasonic assistance.
Catalog No. 50-004-26187 Supplier Medchemexpress LLC Supplier No. HYN93775MG
Platyconic Acid A is an active component of changkil saponins from platycodon grandiflorum and can be used for the research of reducing airway inflammation.
  • Inhibits PMA-induced NF-κB p65 phosphorylation, IκBα phosphorylation and degradation and Akt phosphorylation in a concentration-dependent manner.
  • Decreases PMA-increased MUC5AC mRNA expression in a concentration-dependent manner.
  • Stored at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Soluble in H2O at 25 mg/mL (20.17 mM) with ultrasonic assistance.