Learn More
Medchemexpress LLC HY-N3522 1mg Medchemexpress, Platycoside E CAS:237068-41-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN35221MG
Encompass_Preferred
Medchemexpress, HY-N3522 1mg Platycoside E CAS:237068-41-6 Platycoside E is a platycodigenin-type saponin isolated from the root of Platycodon grandiflorum with haemolytic activity and adjuvant potential. Platycoside E promotes the production of the sera OVA-specific IgG2a and IgG2b antibody in the ovalbumin (OVA)-immunized mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2916