Learn More
Genscript Corporation PLAU/uPA active formBiotin
Catalog No. 5000057675
Encompass
Plasminogen activator urokinase (uPA) is a secreted serine protease whose Dysregulation is often accompanied by various cancers PLAU inhibition could suppress tumor growth Collectively PLAU is necessary for tumor progression and can be a diagnostic and prognostic biomarker in HNSCC
Catalog No. 50-000-57675 Supplier Genscript Corporation Supplier No. Z04587100
Plasminogen activator urokinase (uPA) is a secreted serine protease whose Dysregulation is often accompanied by various cancers PLAU inhibition could suppress tumor growth Collectively PLAU is necessary for tumor progression and can be a diagnostic and prognostic biomarker in HNSCC