Learn More
Medchemexpress LLC PLAU/uPA Human HEK 50ug
SDP
Catalog No. 5000180513
Encompass_Preferred
uPA chain A Protein a key player in the plasminogen activation system selectively cleaves plasminogen initiating fibrinolysis and contributing to tissue remodeling Its precision underscores its role in regulating proteolytic activity emphasizing its significance in the cascade of events leading to fibrinolysis and extracellular matrix remodeling in various physiological processes PLAU/uPA Protein Human (HEK293 His) is the recombinant human-derived PLAU/uPA protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80513 Supplier Medchemexpress LLC Supplier No. HYP7346750UG
uPA chain A Protein a key player in the plasminogen activation system selectively cleaves plasminogen initiating fibrinolysis and contributing to tissue remodeling Its precision underscores its role in regulating proteolytic activity emphasizing its significance in the cascade of events leading to fibrinolysis and extracellular matrix remodeling in various physiological processes PLAU/uPA Protein Human (HEK293 His) is the recombinant human-derived PLAU/uPA protein expressed by HEK293 with C-His labeled tag