Learn More
Medchemexpress LLC PLAU/uPA Mouse Bio 20ug | 20ug
SDP
Catalog No. 5000195299
Encompass_Preferred
PLAU or urokinase-type plasminogen activator (uPA) critically initiates fibrinolysis by cleaving plasminogen to form active plasmin This enzymatic conversion is critical for regulating clot dissolution and tissue remodeling PLAU/uPA Protein Mouse (Biotinylated HEK293 His-Avi) is the recombinant mouse-derived PLAU/uPA protein expressed by HEK293 with C-Avi C-His labeled tag
Catalog No. 50-001-95299 Supplier Medchemexpress LLC Supplier No. HYP7801820UG
PLAU or urokinase-type plasminogen activator (uPA) critically initiates fibrinolysis by cleaving plasminogen to form active plasmin This enzymatic conversion is critical for regulating clot dissolution and tissue remodeling PLAU/uPA Protein Mouse (Biotinylated HEK293 His-Avi) is the recombinant mouse-derived PLAU/uPA protein expressed by HEK293 with C-Avi C-His labeled tag