Learn More
Medchemexpress LLC PLAU/uPA Mouse HEK 100ug | 100ug
SDP
Catalog No. 5000195157
Encompass_Preferred
PLAU or urokinase-type plasminogen activator (uPA) critically initiates fibrinolysis by cleaving plasminogen to form active plasmin This enzymatic conversion is critical for regulating clot dissolution and tissue remodeling PLAU/uPA Protein Mouse (HEK293 C-His-Avi) is the recombinant mouse-derived PLAU/uPA protein expressed by HEK293 with C-His-Avi labeled tag
Catalog No. 50-001-95157 Supplier Medchemexpress LLC Supplier No. HYP78017100UG
PLAU or urokinase-type plasminogen activator (uPA) critically initiates fibrinolysis by cleaving plasminogen to form active plasmin This enzymatic conversion is critical for regulating clot dissolution and tissue remodeling PLAU/uPA Protein Mouse (HEK293 C-His-Avi) is the recombinant mouse-derived PLAU/uPA protein expressed by HEK293 with C-His-Avi labeled tag