Learn More
Medchemexpress LLC PLAU/uPA Rat HEK29 50ug
SDP
Catalog No. 5000180058
Encompass_Preferred
PLAU/uPA proteins lack conserved residues critical for feature annotation propagation suggesting potential differences in molecular interactions in plasminogen activation and fibrinolysis regulation PLAU/uPA Protein Rat (HEK293 His) is the recombinant rat-derived PLAU/uPA protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-80058 Supplier Medchemexpress LLC Supplier No. HYP7356150UG
PLAU/uPA proteins lack conserved residues critical for feature annotation propagation suggesting potential differences in molecular interactions in plasminogen activation and fibrinolysis regulation PLAU/uPA Protein Rat (HEK293 His) is the recombinant rat-derived PLAU/uPA protein expressed by HEK293 with C-His labeled tag