Learn More
ACROBiosystems Recombinant Protein;Human PLAU / uPA Protein;HEK293;50UG;PLU,Urokinase,PLAU,ATF,UPA,URK,u-PA,BDPLT5,QPD
Supplier:  ACROBiosystems PLUH522950UG
Encompass_Preferred
Recombinant Protein;Human PLAU / uPA Protein;50UG;Human PLAU, His Tag (PLU-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Leu 431 (Accession # NP_002649.1).;Human PLAU, His Tag is fused with a polyhistidine tag at the C-terminus, and has a calculated MW of 45.0 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation. Please note that the protein is not activated by protease digestion. In vitro activation is generally recommended for higher activity.;Immobilized Human PLAU, His Tag (Cat. No. PLU-H5229) at 5 μg/mL (100 μL/well) can bind Biotinylated Human uPAR, His Tag, Avi Tag with a linear range of 0.3-2 ng/mL (QC tested).
Catalog No. 50-165-7822