Learn More
Medchemexpress LLC Plazomicin sulfate | 1380078-95-4 | 98.0% | 935.94 g/mol | C25H48N6O10·5/2H2SO4 | 10 MG
SDP
Catalog No. 5000295232
Encompass_Preferred
Plazomicin sulfate is the sulfate salt of plazomicin, an aminoglycoside antibiotic supplied as a solid research reagent for laboratory evaluation of antibacterial activity against multidrug-resistant pathogens. Intended for in vitro research use only, the material is provided as a high-purity powder with specific storage recommendations to preserve stability.
  • High purity suitable for research applications.
  • Solid powder form for accurate weighing and formulation.
  • Defined storage conditions to maintain stability.
  • Small pack sizes suitable for assay development.
  • Applicable to studies of multidrug-resistant Gram-negative bacteria.
Catalog No. 50-002-95232 Supplier Medchemexpress LLC Supplier No. HY16955A10MG
Plazomicin sulfate is the sulfate salt of plazomicin, an aminoglycoside antibiotic supplied as a solid research reagent for laboratory evaluation of antibacterial activity against multidrug-resistant pathogens. Intended for in vitro research use only, the material is provided as a high-purity powder with specific storage recommendations to preserve stability.
  • High purity suitable for research applications.
  • Solid powder form for accurate weighing and formulation.
  • Defined storage conditions to maintain stability.
  • Small pack sizes suitable for assay development.
  • Applicable to studies of multidrug-resistant Gram-negative bacteria.