Learn More
Sino Biological Chinese hamster PLBD2 Protein (His Tag) 100ug
Supplier:  Sino Biological 5A7664A08C100
Encompass_Preferred
A DNA sequence encoding the chinese hamster PLBD2 (G3I6T1-1) (Leu38-Asp585) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6506