Learn More
Cayman Chemical PLC thioPIP2 sodium salt
SDP
Supplier:  Cayman Chemical 10007839100 UG
Encompass_Preferred
A novel, chromogenic substrate for PLC; hydrolysis by PLC yields a free thiol which reacts with chromogenic reagents such as DTNB (Ellman’s reagent) to allow quantitation of PLC activity
Catalog No. 50-288-8213