Learn More
Cayman Chemical PLC thioPIP2 sodium salt
SDP
Catalog No. 502889104
Encompass_Preferred
A novel, chromogenic substrate for PLC; hydrolysis by PLC yields a free thiol which reacts with chromogenic reagents such as DTNB (Ellman’s reagent) to allow quantitation of PLC activity
Catalog No. 50-288-9104 Supplier Cayman Chemical Supplier No. 10007839500 UG
A novel, chromogenic substrate for PLC; hydrolysis by PLC yields a free thiol which reacts with chromogenic reagents such as DTNB (Ellman’s reagent) to allow quantitation of PLC activity