Learn More
Cayman Chemical PLC thio-PIP2 (sodium salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502888014
Encompass_Preferred
A novel, chromogenic substrate for PLC; hydrolysis by PLC yields a free thiol which reacts with chromogenic reagents such as DTNB (Ellman’s reagent) to allow quantitation of PLC activity
Catalog No. 50-288-8014 Supplier Cayman Chemical Supplier No. 100078391 MG
A novel, chromogenic substrate for PLC; hydrolysis by PLC yields a free thiol which reacts with chromogenic reagents such as DTNB (Ellman’s reagent) to allow quantitation of PLC activity