Learn More
Aviva Systems Biology Corp PLCD1 Antibody - N-terminal region (ARP56690_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56690_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLCD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLCD1. Alternate names: NDNC3, PLC-IIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61954