Learn More
Akina Inc Poly(lactide-co-caprolactone)-b-Poly(ethylene glycol)-b-Poly(lactide-co-caprolactone) (per g)
SDP
Catalog No. NC1818694
Encompass
PLCL-PEG-PLCL (Mw ~1700:1500:1700 Da) CL:LA 90:10 Poly(lactide-co-caprolactone)-b-Poly(ethylene glycol)-b-Poly(lactide-co-caprolactone) (per g)
Catalog No. NC1818694 Supplier Akina Inc Supplier No. AK1391G
PLCL-PEG-PLCL (Mw ~1700:1500:1700 Da) CL:LA 90:10 Poly(lactide-co-caprolactone)-b-Poly(ethylene glycol)-b-Poly(lactide-co-caprolactone) (per g)