Learn More
Innovative Research Inc Pld Hmn Breast Milk Lyo 100mg
Catalog No. 502744039
Encompass_Preferred
Pooled Human Breast Milk Lyophilized 100mg - Pooled Human Breast Milk Lyophilized from Innovative Research is collected, pooled, and then lyophilized. Breast milk contains complex proteins, lipids, carbohydrates, and other biologically-active components.
Catalog No. 50-274-4039 Supplier Innovative Research Inc Supplier No. IRHUBMKLY100MG
Pooled Human Breast Milk Lyophilized 100mg - Pooled Human Breast Milk Lyophilized from Innovative Research is collected, pooled, and then lyophilized. Breast milk contains complex proteins, lipids, carbohydrates, and other biologically-active components.