Learn More
Innovative Research Inc Pld Hmn Comp Srm 1ml
Supplier:  Innovative Research Inc ICSER1ML
Encompass_Preferred
Pooled Human Complement Serum 1ml - Human Complement Serum is a complex mix of serum proteins which are activated sequentially when antibodies in blood with immunity interact with the corresponding antigens. Complement proteins will quickly degrade if the serum is not prepared and stored correctly. Our complement-preserved serum is freshly collected aseptically and kept cold, and then flash-frozen to prevent degradation of complement activity. Please note that this product does not allow for donor requests off-the-shelf, but we can create custom pooled lots to your exact specifications upon request. Contact us for details & pricing. Each unit is tested for the standard FDA-required viral markers using FDA-approved methods. While we make every effort to ensure the safety of our products, we recommend following universal precautions as laid out the following organizations WHO, CDC and OSHA, handle biological materials as if capable of spreading infectious disease
Catalog No. 50-274-4600