Learn More
Innovative Research Inc Pld Hmn Semen 20ml
Catalog No. 502744032
Encompass_Preferred
Pooled Human Semen 20ml - Pooled Human Semen from Innovative Research is collected from two or more unique donors in an FDA-approved facility, and processed using ISO-approved procedures. During the process of ejaculation, sperm passes through the ejaculatory ducts and mixes with fluids from the seminal vesicles, the prostate, and the bulbourethral glands to form the semen. The seminal vesicles produce a yellowish viscous fluid rich in fructose and other substances that makes up about 70% of human semen. The prostatic secretion, influenced by dihydrotestosterone, is a whitish (sometimes clear), thin fluid containing proteolytic enzymes, citric acid, acid phosphatase and lipids.
Catalog No. 50-274-4032 Supplier Innovative Research Inc Supplier No. IRHUSM20ML
Pooled Human Semen 20ml - Pooled Human Semen from Innovative Research is collected from two or more unique donors in an FDA-approved facility, and processed using ISO-approved procedures. During the process of ejaculation, sperm passes through the ejaculatory ducts and mixes with fluids from the seminal vesicles, the prostate, and the bulbourethral glands to form the semen. The seminal vesicles produce a yellowish viscous fluid rich in fructose and other substances that makes up about 70% of human semen. The prostatic secretion, influenced by dihydrotestosterone, is a whitish (sometimes clear), thin fluid containing proteolytic enzymes, citric acid, acid phosphatase and lipids.