Learn More
Innovative Research Inc Pld Hmn Srm Off Clot Fml/ Ml
Supplier:  Innovative Research Inc ISERFM100ML
Encompass_Preferred
Pooled Human Serum Off The Clot Female and Male Mix - Pooled Human Serum from Innovative Research is whole blood derived. The whole blood is collected from donors in an FDA-approved collection center. The blood is allowed to clot and is then processed into serum via centrifugation, and is frozen immediately after processing.
Catalog No. 50-274-3986