Learn More
Innovative Research Inc Pld Hmn Sweat 1ml
Catalog No. 502744024
Encompass_Preferred
Pooled Human Sweat 1ml - Pooled Human Sweat from Innovative Research is collected from three or more unique donors per lot, pooled, and stored frozen.
Catalog No. 50-274-4024 Supplier Innovative Research Inc Supplier No. IRHUSW1ML
Pooled Human Sweat 1ml - Pooled Human Sweat from Innovative Research is collected from three or more unique donors per lot, pooled, and stored frozen.