Learn More
Innovative Research Inc Pld Hmn Urn 50ml
Catalog No. 502744020
Encompass_Preferred
Pooled Human Urine 50ml - Pooled Human Urine from Innovative Research is collected fresh from multiple normal donors. The urine is collected and pooled from two or more donors immediately after collection from the final donor. The material is then stored and shipped frozen, however, upon request, we can supply pooled human urine fresh, never frozen.
Catalog No. 50-274-4020 Supplier Innovative Research Inc Supplier No. IRHUURE50ML
Pooled Human Urine 50ml - Pooled Human Urine from Innovative Research is collected fresh from multiple normal donors. The urine is collected and pooled from two or more donors immediately after collection from the final donor. The material is then stored and shipped frozen, however, upon request, we can supply pooled human urine fresh, never frozen.