Learn More
Aviva Systems Biology Corp PLD3 Antibody - N-terminal region (ARP44768_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44768_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLD3. Alternate names: HU-K4, HUK4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53435