Learn More
Biomatik Corporation Pld4 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11897
Encompass_Preferred
Pld4 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PLD4. The Pld4 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Phospholipase D4 Protein (52-506Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PLD4. Accession Number: Q96BZ4. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9231