Learn More
Aviva Systems Biology Corp PLDN Antibody - N-terminal region (ARP54889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLDN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLDN. Alternate names: PA, PALLID, HPS9, PLDNApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61355