Learn More
Bachem (Des-Ser3)-ACTH (1-24) (human, bovine, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4059984.0005
Encompass_Preferred
(Des-Ser3)-ACTH (1-24) (human, bovine, rat) Trifluoroacetate, H-Ser-Tyr-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH, 1926163-12-3, C133H205N39O29S, Mr 2846.40, 5mg. (Des-Ser3)-Tetracosactide. Impurity of tetracosactide, which is a synthetic peptide analog of the human adrenocorticotropic hormone that stimulates the production of cortisol.
Catalog No. 50-194-6985