Learn More
Medchemexpress LLC HY-108741A 10mg Medchemexpress, Plecanatide acetate CAS:1075732-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108741A10MG
Encompass_Preferred
Medchemexpress, HY-108741A 10mg Plecanatide acetate CAS:1075732-84-1 Plecanatide acetate is a guanylate cyclase-C (GC-C) receptor agonist, with an EC50 of 190 nM in T84 cells. Plecanatide acetate shows anti-inflammatory activity in models of murine colitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8624