Learn More
Medchemexpress LLC Pleconaril | 153168-05-9 | 99.48% | 381.35 | 200 MG
SDP
Catalog No. 5000401571
Encompass_Preferred
Pleconaril is a picornavirus capsid binding inhibitor that prevents attachment and/or virus uncoating. It has potential for use in research related to rhinoviruses and enteroviruses.
  • Prevents attachment and/or virus uncoating
  • For research use only
Catalog No. 50-004-01571 Supplier Medchemexpress LLC Supplier No. HY19952200MG
Pleconaril is a picornavirus capsid binding inhibitor that prevents attachment and/or virus uncoating. It has potential for use in research related to rhinoviruses and enteroviruses.
  • Prevents attachment and/or virus uncoating
  • For research use only