Learn More
MSC SC Johnson Multi-Surface Wet Wipes, Fresh Citrus, 25 Sheets
Catalog No. 502867959
Encompass
These pre-saturated, flat sheet multi-surface cleaner wet wipes are designed for general purpose cleaning on hard surfaces. They provide effective cleaning with a fresh citrus scent and are suitable for diverse professional environments.
  • Pre-saturated flat sheet wipes
  • Designed for multi-surface cleaning tasks
  • Measures 7 in x 10 in per sheet
  • Features a fresh citrus scent
  • Comes in a resealable pack for convenience
  • Applicable in hospitality, education, industrial, and office settings
  • Exhibits a pH of 6.0 at 20 °C
  • Each canister contains 25 sheets
Catalog No. 50-286-7959 Supplier MSC Supplier No. 96149273
These pre-saturated, flat sheet multi-surface cleaner wet wipes are designed for general purpose cleaning on hard surfaces. They provide effective cleaning with a fresh citrus scent and are suitable for diverse professional environments.
  • Pre-saturated flat sheet wipes
  • Designed for multi-surface cleaning tasks
  • Measures 7 in x 10 in per sheet
  • Features a fresh citrus scent
  • Comes in a resealable pack for convenience
  • Applicable in hospitality, education, industrial, and office settings
  • Exhibits a pH of 6.0 at 20 °C
  • Each canister contains 25 sheets