Learn More
New England Biolabs, Inc. PleI – 1000 units
SDP
Catalog No. 50812164
Encompass
A restriction endonuclease that recognizes the sequence GAGTCNNNN^N_.
Catalog No. 50-812-164 Supplier New England Biolabs, Inc. Supplier No. R0515S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GAGTCNNNN^N_.