Learn More
Sigma Aldrich Fine Chemicals Biosciences Pleiotrophin human 97 SDS
Catalog No. 502904194
Encompass
Pleiotrophin (PTN) is a multifunctional growth factor. The protein coded by the PTN gene is a member of the heparin-binding cytokines family. This gene is located on human chromosome 7. Pleiotrophin is also known as heparin-binding brain mitogen (HBBM) heparin-binding growth factor-8 (HBGf-8) heparin-binding growth-associated molecule (HB-GAM) and osteoblast-specific factor (OSf-1). This gene is highly expressed in brain uterus gut muscle lung and skin. Pleiotrophin mRNA is expressed in osteoblasts chondrocytes tumor cells fibroblasts astrocytes and Schwann cells.
Catalog No. 50-290-4194 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P533350UG
Pleiotrophin (PTN) is a multifunctional growth factor. The protein coded by the PTN gene is a member of the heparin-binding cytokines family. This gene is located on human chromosome 7. Pleiotrophin is also known as heparin-binding brain mitogen (HBBM) heparin-binding growth factor-8 (HBGf-8) heparin-binding growth-associated molecule (HB-GAM) and osteoblast-specific factor (OSf-1). This gene is highly expressed in brain uterus gut muscle lung and skin. Pleiotrophin mRNA is expressed in osteoblasts chondrocytes tumor cells fibroblasts astrocytes and Schwann cells.