Learn More
Aviva Systems Biology Corp PLEKHA4 Antibody - N-terminal region (ARP42572_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42572_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLEKHA4. Alternate names: PEPP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58271