Learn More
Aviva Systems Biology Corp PLEKHA7 Antibody - middle region (ARP53316_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53316_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHA7. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PLEKHA7. Alternate names: DKFZp686M22243ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-825