Learn More
Aviva Systems Biology Corp PLEKHB2 Antibody - middle region (ARP56309_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56309_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLEKHB2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLEKHB2. Alternate names: EVT2, FLJ20783, FLJ23679, FLJ30436ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61804