Learn More
Aviva Systems Biology Corp Plekhf2 Antibody - N-terminal region (ARP50680_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50680_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Plekhf2. It was validated on Western Blot. . Alternate names: 1110070J07Rik, AA673237, ZFYVE18ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-525